Cohesive Pocket Samplers
About this item
The cohesive pocket sampler is designed for taking discreet samples of cohesive powders.
- Ingenious scraper pushes powder into containment when rotated
- Made from 316L stainless steel
- Available from stock
- Fast delivery
The cohesive pocket sampler is perfect for taking discreet samples of powders with poor flow characteristics. The scraper pushes powder into the sampler when rotated, while a solid inner rod ensures that samples are kept separate from each other.
3 Options Available Below
Product Details & Documents
The cohesive pocket sampler is designed for taking discreet samples of cohesive powders.
- Ingenious scraper pushes powder into containment when rotated
- Made from 316L stainless steel
- Available from stock
- Fast delivery
The cohesive pocket sampler is perfect for taking discreet samples of powders with poor flow characteristics. The scraper pushes powder into the sampler when rotated, while a solid inner rod ensures that samples are kept separate from each other.
Product Family Options
Product Information
- MaterialLengthVolumeAvailabilityPrice
- SAMP2220B-600Cohesive pocket sampler
- SAMP2220B-900Cohesive pocket sampler
- SAMP2220B-1500Cohesive pocket sampler