Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Protein Extraction
Protein Extraction Kits
Tryptic Digest of Bovine Serum Albumin
Tryptic Digest of Bovine Serum Albumin
Tryptic Digest of Bovine Serum Albumin
Catalog # BRUK8217498
Supplier:
BRUKER DALTRONIK
New Product
CAS Number:
Expand image
Tryptic Digest of Bovine Serum Albumin
Catalog # BRUK8217498
Supplier:
BRUKER DALTRONIK
New Product
CAS Number:
Specifications
Pk:
1
Description:
Tryptic digest of bovine serum albumin, for 5×250 spots
Specifications
About this item
Product Details
Product Details