Order Entry
Germany
ContactUsLinkComponent
 
MTT Formazan, powder
  M2003-1G
 :  
undefined
MTT Formazan, powder
  M2003-1G
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance (Form):
    Powder
  • Carbon:
    62.8 - 66.1 %
  • Nitrogen:
    20.4 - 21.4 %
  • Purity (TLC):
    ≥ 97 %
  • Solubility (Colour):
    Conforms to Requirements

 

 

 :
 :335.43 g/mol
 :M-Clarity™ quality level = MQ200
 :Room Temperature
 :57360-69-7
 :C₁₈H₁₇N₅S
 :MFCD00080731