Orders LinkContactUsLinkComponent
STEMdiff™ Cardiomyocyte Support Medium
STEMdiff™ Cardiomyocyte Support Medium
STEMdiff™ Cardiomyocyte support medium, 250 mL
Catalog # STMC05027
Marketsource
STEMdiff™ Cardiomyocyte Support Medium
Catalog # STMC05027
Marketsource
CAS Number:  
STEMdiff™ Cardiomyocyte Support Medium
STEMdiff™ Cardiomyocyte Support Medium
STEMdiff™ Cardiomyocyte support medium, 250 mL
CAS Number:  
Price
200,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Medium for thawing and culturing hPSC-derived cardiomyocytes.
STEMdiff™ Cardiomyocyte Support Medium is a versatile medium that limits stress on human pluripotent stem cell (hPSC)-derived cardiomyocytes when transitioning from cryopreservation to thawing and from harvesting to replating of hPSC-derived cardiomyocytes. After thawing or replating, the functional capacity of hPSC-derived cardiomyocytes is retained and cells can be used in various downstream applications and analyses. The hPSC-derived cardiomyocytes can be further maintained long-term using STEMdiff™ Cardiomyocyte Maintenance Kit (Catalog #05020).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    250 mL
  • Description:
    STEMdiff™ Cardiomyocyte support medium
Specifications