ContactUsLinkComponent
MethoCult™ M3234 Medium
MethoCult™ M3234 Medium
Catalog # STMC03234
CAS Number:  
undefined
MethoCult™ M3234 Medium
Catalog # STMC03234
CAS Number:  
Specifications
  • Pk:
    80 mL
  • Description:
    MethoCult™ M3234
Specifications
About this item
Methylcellulose-based medium without cytokines (including EPO) for mouse cells.
MethoCult™ M3234 is an incomplete medium formulation that contains methylcellulose in Iscove's MDM. MethoCult™ M3234 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver, when the appropriate growth factors and supplements are added. This formulation does not contain erythropoietin or other cytokines.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').