ContactUsLinkComponent
NaïveCult™ Induction Kit for the generation of reset hPSCs
NaïveCult™ Induction Kit for the generation of reset hPSCs
Catalog # STMC05580
CAS Number:  
undefined
NaïveCult™ Induction Kit for the generation of reset hPSCs
Catalog # STMC05580
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    NaïveCult™ Induction kit for the generation of reset hPSCs
Specifications
About this item
Defined medium kit for transgene-free induction of reset naïve human pluripotent stem cells - 1 Kit
Perform transgene-free chemical induction of reset human naïve embryonic stem (ES) and induced pluripotent stem (iPS) cells from the primed state using a defined cell culture medium. Reset naïve human pluripotent stem cells (hPSCs) generated using this kit demonstrate uniformly domed, phase-bright colonies with refractive edges and maintain gene expression profiles associated with reset naïve hPSCs. Based on the formulation developed under license from the cambridge stem cell institute at the university of cambridge, UK, this kit works robustly across multiple primed human ES and iPS cells maintained in mTeSR™1 or TeSR™-E8™.
Note that NaïveCult™ induction kit requires a histone deacetylase inhibitor (HDACi; either sodium butyrate or valproic acid) that is sold separately.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').