ContactUsLinkComponent
MethoCult™ H4230 Medium
MethoCult™ H4230 Medium
Catalog # STMC04230
CAS Number:  
undefined
MethoCult™ H4230 Medium
Catalog # STMC04230
CAS Number:  
Specifications
  • Pk:
    80 mL
  • Description:
    MethoCult™ H4230
Specifications
About this item
Methylcellulose-based medium without cytokines for human cells.
MethoCult™ H4230 is an incomplete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilised peripheral blood, peripheral blood, and cord blood samples. This formulation does not contain erythropoietin (EPO) or other cytokines, allowing the addition of cytokines to meet the specific requirements of investigators.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').