ClonaCell™-HY Medium E
Product Details & Documents
Hybridoma growth medium with hypoxanthine and thymidine (serum-containing).
ClonaCell™-HY medium E is a serum-containing, nutritionally rich liquid medium optimised for hybridoma expansion after hypoxanthine, aminopterin, thymidine (HAT) selection. The medium contains hypoxanthine and thymidine (HT) and is used to wean hybridomas off aminopterin used during the selection process. This medium has been verified for use in mouse and rat hybridoma development and monoclonal antibody production and reportedly is compatible for production, cloning, and expansion of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster. ClonaCell™-HY medium E has proven effective for subcloning and stabilisation of unstable hybridoma cell lines.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
- VE:500 mL
- Bezeichnung:ClonaCell™-HY medium E