ContactUsLinkComponent
ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator
ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator
  STMC10990
 :  
undefined
ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator
  STMC10990
 :  
  • Pk:
    1 KIT
  • Description:
    ImmunoCult™ human CD3/CD28/CD2 T cell activator
cGMP, human T cell activation and expansion reagent.
Achieve robust activation and expansion of human T cells for use in clinical applications - without the use of magnetic beads, feeder cells, or antigens. This product's gentle activation stimulus ensures a high viability of activated T cells, which can be further expanded in ImmunoCult™-XF, a high-performance T cell expansion medium manufactured under relevant cGMP regulations and guidelines.
ImmunoCult™ Human CD3/CD28/CD2 T cell activator consists of soluble antibody complexes that bind to and cross-link CD3, CD28, and CD2 cell surface ligands, providing the required primary and co-stimulatory signals for T cell culture and activation. ImmunoCult™ Human CD3/CD28/CD2 T cell activator is designed for cell therapy clinical research applications, qualified for use as an ancillary material (AM) following the framework outlined in USP 1043 and/or PH. EUR. 5.2.12. STEMCELL can work with you to qualify this reagent as an AM under an approved Investigational New Drug (IND) application, Biological Licensing Application (BLA), or Clinical Trial Application (CTA).
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').