Order Entry
Germany
ContactUsLinkComponent
PolTech Ultra DES Disinfectant Cleaners
PolTech Ultra DES Disinfectant Cleaners
Catalog # BSNC2600044
Supplier:  Basan
New Product
PolTech Ultra DES Disinfectant Cleaners
Catalog # BSNC2600044
Supplier:  Basan
New Product

Specifications

  • Capacity:
    6×1 L
  • Description:
    PolTech Ultra DES disinfectant cleaner
  • Seq:
    02
  • Pk:
    1
  • pH value:
    pH in solution of 0,25%: 9 ±0,5, Pure pH: 11 ±0,5

Specifications

About this item

Disinfectant cleaner for floors and surfaces in medical settings.

PolTech Ultra DES (bactericidal, yeasticidal and virucidal against enveloped viruses) allows the cleaning and disinfection of floors and all washable surfaces (tables, worktops, railings, etc.) in a single operation.

Particularly suitable for disinfecting surfaces in medical settings and in sectors such as public buildings, the food and non-food industry or the professional kitchen. Product approved for food contact.

Certifications: Bactericidal: EN1276, EN13697, Bactericide in medical settings: EN13727, Yeasticide: EN1650, EN13697, Yeasticide in medical settings: EN13624, virucidal against enveloped viruses, adenovirus and coronavirus: EN14476.