Order Entry
Germany
ContactUsLinkComponent
Gram's decolourising solution for Gram staining, Sigma-Aldrich®
Flammable
Irritant
Gram's decolourising solution for Gram staining, Sigma-Aldrich®
Certificates
Flammable
Irritant
Gram's decolourising solution for Gram staining, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (colour):
    Colourless
  • Appearance (form):
    Liquid
  • Density D20/4:
    0.785 - 0.814
  • Refractive index N20/D:
    1.356 - 1.366
  • Proton NMR spectrum:
    Conforms to structure

Specification Test Results

Related Information