ESD Safety Ankle Boots, Lacing Sytem, Stefano XXSG Pro Boa® GTX Mid S3
Supplier: ELTEN
New Product
Certificates
About this item
ESD safety ankle boot made from cowhide leather upper and a PU/PU sole (Wellmaxx Safety Grip).
- All weather Gore-Tex performance membrane
- Full, padded bellows tongue
- Full-length inlay sole ARTISAN ESD
- Boa® fit lacing system
- Non-metallic, penetration-resistant midsole
- PU toe protection
- Steel toe cap
Suitable for areas where exposure to moisture is expected (S2). Areas where there is a risk of penetration from pointed and sharp objects (S3). Areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black/blue
Certifications: EN ISO 20345 S3 CI/SRC, form B.
Specifications
- Catalog No:
- ELTN768751/47
- ELTN768751/36
- ELTN768751/37
- ELTN768751/38
- ELTN768751/42
- ELTN768751/39
- ELTN768751/43
- ELTN768751/40
- ELTN768751/44
- ELTN768751/41
- ELTN768751/45
- ELTN768751/46
- ELTN768751/48
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001