ESD Safety Ankle Boots, Lace-Up, Jo_Champ Mid S3
Supplier: ELTEN
New Product
Certificates
About this item
ESD safety ankle boot made from cowhide leather upper and a PU/PU sole (jo_EXPLORE).
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Soft fleece ESD insole
- Full-length inlay sole (jo_TITAN ESD)
- Steel midsole
- Steel toe cap
- PU scuff cap
Suitable for indoors and outdoors, areas where exposure to moisture is expected (S2), where there is a risk of penetration from pointed and sharp objects (S3) and/or where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black
Certifications: EN ISO 20345 S3 FO/LG/SC/SR, form B.
Specifications
- Catalog No:
- ELTN16421/48
- ELTN16421/39
- ELTN16421/40
- ELTN16421/41
- ELTN16421/42
- ELTN16421/43
- ELTN16421/44
- ELTN16421/45
- ELTN16421/36
- ELTN16421/46
- ELTN16421/47
- ELTN16421/37
- ELTN16421/38
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001