Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet 450)
Supplier: AAT BIOQUEST
Certificates
About this item
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: mFluor™ Violet 450
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Specifications
- Catalog No:
- AATB103400Z0
- AATB103400Z1
- Antigen symbol:
- CD34
- CD34
- Antigen name:
- CD34 antigen
- CD34 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ Violet 450
- mFluor™ Violet 450
- Clone:
- 4H11
- 4H11
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20