Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 840)
Supplier: AAT BIOQUEST
Certificates
About this item
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 840
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB101920Q1
- AATB101920Q0
- Antigen symbol:
- CD19
- CD19
- Antigen name:
- CD19 antigen
- CD19 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 840
- iFluor® 840
- Clone:
- HIB19
- HIB19
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20