Order Entry
Germany
ContactUsLinkComponent
Anti-ATF4 Rabbit Polyclonal Antibody
Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # PRSI56-296
Supplier:  ProSci Inc.
New Product
undefined
Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # PRSI56-296
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclic AMP-dependent transcription factor ATF-4
  • Antigen symbol:
    ATF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Amino Acid Range = 171-198
  • Form:
    Liquid
  • Gene ID:
    P18848
  • Antigen synonyms:
    ATF4
  • Modification:
    None
  • Storage buffer:
    Supplied in PBS with 0,09% (W/V) sodium azide
  • Molecular weight:
    39 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Shipping temperature:
    4 °C
  • Immunogen:
    This ATF4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 171-198 amino acids from the Central region of human ATF4.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification.
  • Size:
    400 µl
  • Pk:
    400 µl

Specifications

About this item

This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication.

  • For WB starting dilution is: 1:1000

Type: Primary
Antigen: ATF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human