Specifications
- Antibody type:Primary
- Antigen name:AMP deaminase 3
- Antigen symbol:AMPD3
- Clonality:Monoclonal
- Clone:ETAD3-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q01432
- Antigen synonyms:AMPD3
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant full-length human protein was used as the immunogen for the AMPD3 antibody.
- Tested applications:Immunocytochemistry
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
AMPD3 is a member of the AMP deaminase gene family. It is a highly regulated enzyme that catalyses the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. This gene encodes the erythrocyte (E) isoforms, whereas other family members encode isoforms that predominate in muscle (M) and liver (L) cells. Mutations in the gene lead to the clinically asymptomatic, autosomal recessive condition erythrocyte AMP deaminase deficiency. [RefSeq].
Immunofluorescence: 0,5 - 1 µg/ml Immunocytochemistry: 1 - 2 µg/ml for 30 min at RT Immunohistology (FFPE): 2-4 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the AMPD3 antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 min
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: AMPD3
Clonality: Monoclonal
Clone: ETAD3-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human