- Antibody type:Primary
- Antigen name:Thymidine phosphorylase
- Antigen symbol:TYMP
- Clonality:Monoclonal
- Clone:P-GF.44C
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P19971
- Antigen synonyms:TYMP
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Human recombinant full length protein was used as the immunogen for this Thymidine Phosphorylase antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Recognises a 485 amino acid protein (55 kDa monomer/110 kDa homodimer), identified as platelet-derived endothelial growth factor (PD-ECGF), also called Thymidine Phosphorylase (TP, Tymp) or Gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. Its enzymatic activity is crucial for angiogenic activity (metabolite is angiogenic). Higher levels of serum TP/PD-ECGF are observed in cancer patients. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites (e.g. 5 -deoxy-5-fluorouridine to 5-FU). High intra-cellular levels of TP/PD-ECGF are associated with increased chemosensitivity to such antimetabolites.
IP: 1-2µg/500µg lysate IHC (FFPE): 1 - 2 µg/ml for 30 min at RT WB: 0,5 - 1 µg/ml The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Thymidine Phosphorylase antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: TYMP
Clonality: Monoclonal
Clone: P-GF.44C
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat