Anti-GRIN2A Rabbit Polyclonal Antibody
PRSI15-236
- Antibody type:Primary
- Antigen name:Glutamate receptor ionotropic, NMDA 2A
- Antigen symbol:GRIN2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:2903
- Antigen synonyms:NR2A|hNR2A|Glutamate receptor ionotropic|N-methyl D-aspartate receptor subtype 2A|GRIN2A|NMDA 2A|GluN2A|NMDAR2A|Glutamate [NMDA] receptor subunit epsilon-1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 23-120 of human GRIN2A (NP_000824.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: GRIN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat