Anti-UBE2V2 Rabbit Polyclonal Antibody
Catalog # PRSI22-619
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating enzyme E2 variant 2
- Antigen symbol:UBE2V2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:7336
- Antigen synonyms:S. cerevisiae|UEV2|DDVit-1|EDPF-1|DDVIT1|1 alpha|UBE2V2|EDPF1|MMS2|ubiquitin-conjugating enzyme E2 variant 2|25-dihydroxyvitamin D3-inducible|EDAF-1|UEV-2|methyl methanesulfonate sensitive 2|enterocyte differentiation promoting factor|homolog of
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human UBE2V2 (NP_003341.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: UBE2V2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat