Anti-DOCK7 Rabbit Polyclonal Antibody
Catalog # PRSI16-891
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Dedicator of cytokinesis 7
- Antigen symbol:DOCK7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:85440
- Antigen synonyms:dedicator of cytokinesis protein 7|ZIR2|EIEE23
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human DOCK7
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DOCK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat