Order Entry
Germany
ContactUsLinkComponent
 
Anti-CREB3L3 Rabbit Polyclonal Antibody
  PRSI16-697
New Product
Anti-CREB3L3 Rabbit Polyclonal Antibody
  PRSI16-697
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CAMP responsive element binding protein 3 like 3
  • Antigen symbol:
    CREB3L3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    84699
  • Antigen synonyms:
    cAMP-responsive element-binding protein|CREB-H|CREB/ATF family transcription factor|CREBH|HYST1481|cyclic AMP-responsive element-binding protein 3-like protein 3|hepatic-specific
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 371-460 of human CREB3L3 (NP_001258924,1)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: CREB3L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat