Anti-LIPA Rabbit Polyclonal Antibody
Catalog # PRSI22-177
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Lipase A, lysosomal acid, cholesterol esterase
- Antigen symbol:LIPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:3988
- Antigen synonyms:Acid cholesteryl ester hydrolase|CESD|LAL|Cholesteryl esterase
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 110-399 of human LIPA (NP_001121077.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: LIPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat