Anti-CCT6A Rabbit Polyclonal Antibody
PRSI19-097
New Product
- Antibody type:Primary
- Antigen name:Chaperonin containing TCP1 subunit 6A
- Antigen symbol:CCT6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:908
- Antigen synonyms:CCT-zeta-1|CCT-zeta|T-complex protein 1|TCP-1-zeta|acute morphine dependence related protein 2|CCT6|chaperonin containing T-complex subunit 6|histidine transport regulator 3|HTR3|zeta subunit|MoDP-2|chaperonin containing TCP1|subunit 6A (zeta 1)|amino acid transport defect-complementing|TTCP2T-complex protein 1 subunit zeta|Cctz|TCP2TCPZ
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 80-250 of human CCT6A (NP_001753.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CCT6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat