Order Entry
Germany
ContactUsLinkComponent
 
Anti-KMT5C Recombinant Antibody [Clone: RAB-C409]
Catalog # PRSI24-860
Supplier:  ProSci Inc.
Anti-KMT5C Recombinant Antibody [Clone: RAB-C409]
Catalog # PRSI24-860
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    lysine methyltransferase 5C
  • Antigen symbol:
    KMT5C
  • Clonality:
    Recombinant
  • Clone:
    RAB-C409
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    84787
  • Antigen synonyms:
    Histone-lysine N-methyltransferase KMT5C|anti-SUV420H2-RAB-C409|Suppressor of variegation 4-20 homolog 2|Suv4-20h2|Lysine-specific methyltransferase 5C|48G1_11|2.1.1.361|Lysine N-methyltransferase 5C|2.1.1.362|Su(var)4-20 homolog 2|[histone H4]-N-methyl-L-lysine20 N-methyltransferase KMT5B|[histone H4]-lysine20 N-methyltransferase KMT5B
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was obtained by recombinant antibody (rAb) phage display recognizing SUV420H2 protein under non-denaturing conditions
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: lysine methyltransferase 5C
Clonality: Recombinant
Clone: RAB-C409
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human