Anti-L3MBTL1 Recombinant Antibody [Clone: RAB-C213]
Catalog # PRSI24-846
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:L3MBTL histone methyl-lysine binding protein 1
- Antigen symbol:L3MBTL1
- Clonality:Recombinant
- Clone:RAB-C213
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:26013
- Antigen synonyms:H-l(3)mbt|anti-L3MBTL1-RAB-C213|L(3)mbt protein homolog|L(3)mbt-like|L3MBTL1-A003-1453|L3MBTL1|Lethal(3)malignant brain tumor-like protein 1|Lethal 3 malignant brain tumor-like protein 1|H-l(3)mbt protein
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing L3MBTL1 protein under non-denaturing conditions
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: L3MBTL histone methyl-lysine binding protein 1
Clonality: Recombinant
Clone: RAB-C213
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human