Anti-TYRP1 Recombinant Antibody [Clone: TA99]
Catalog # PRSI24-232
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:tyrosinase related protein 1
- Antigen symbol:TYRP1
- Clonality:Recombinant
- Clone:TA99
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:7306
- Antigen synonyms:Tyrosinase-related protein 1|Catalase B|5,6-dihydroxyindole-2-carboxylic acid oxidase|Glycoprotein 75|TRP|DHICA oxidase|TRP-1|Melanoma antigen gp75|TRP1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:70-75 kDa pigmentation-associated glycoprotein in human melanoma cell lines
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: tyrosinase related protein 1
Clonality: Recombinant
Clone: TA99
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human