Anti-Ccr7 Recombinant Antibody [Clone: 4B12]
Catalog # PRSI24-799
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) receptor 7
- Antigen symbol:Ccr7
- Clonality:Recombinant
- Clone:4B12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:12775
- Antigen synonyms:MIP-3 beta receptor|Epstein-Barr virus-induced G-protein coupled receptor 1|EBV-induced G-protein coupled receptor 1|EBI1|C-C CKR-7|CD197|CC-CKR-7|C-C chemokine receptor type 7
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing rats with mouse CCR7 antigen
- Tested applications:Functional Assays
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: chemokine (C-C motif) receptor 7
Clonality: Recombinant
Clone: 4B12
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse