Orders LinkContactUsLinkComponent
Dishes, disposable
Dishes, disposable
Supplier:   BOCHEM
Dishes, disposable
Supplier:  BOCHEM
Certificates
Dishes, disposable
Dishes, disposable
4 Options Available Below
Product Details & Documents
Aluminium.
  • Flat bottom and crimped sides for rigidity
  • Oil-free
  • Slightly tapered for easy stacking
Sturdy, disposable dishes feature crimped sides. Ideal for general-purpose weighing, dispensing, and storage applications. Dishes also function as evaporating dishes, parts containers, or dust covers.
Product Family Options
Product Information
  • Capacity
    Int.Ø×H
    Ext.Ø
    Height
    Availability
    Price
  • 250 ml
    114×50 mm
    114 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 66,50 €
    66,50 €
    1 * 100 ST
       66,50 €
  • 28 ml
    64×13 mm
    64 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 33,60 €
    33,60 €
    1 * 100 ST
       33,60 €
  • 110 ml
    99×25 mm
    99 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 35,90 €
    35,90 €
    1 * 100 ST
       35,90 €
  • 125 ml
    96×24 mm
    96 mm
    24 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,60 €
    48,60 €
    1 * 100 ST
       48,60 €