Turpentine oil
Catalog # CSLOG184-5L
Supplier: CAESAR LORENTZ
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
About this item
:
Boiling Pt:153…175 °C (1013 hPa)
Melting Pt:–60…–50 °C
Density:0.854…0.868 g/cm³ (25 °C)
Flash Pt:35 °C
EINECS:232-350-7
UN:1299
ADR:3,III
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:8006-64-2
MDL Number:MFCD00147790