Pyridinium chlorochromate
Catalog # MOLEM29524115
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:215.56 g/mol
Merck Index:13,08064
Melting Pt:205 °C
EINECS:247-595-5
UN:1479
ADR:5.1,II
CAS Number:26299-14-9
Formula:C₅H₆ClCrNO₃
MDL Number:MFCD00013106