Order Entry
Germany
ContactUsLinkComponent
 
 
 
Phenolphthalein 0.5 wt. % in ethanol: water (1:1), Sigma-Aldrich®
 
undefined
 
 
Phenolphthalein 0.5 wt. % in ethanol: water (1:1), Sigma-Aldrich®
 

 

  • Appearance (Colour):
    Colourless
  • Appearance (Form):
    Liquid
  • Visual Interval
    pH 8.0 Colorless:
    Pass
  • Visual Interval
    pH 8.2 Pale Pink:
    Pass
  • Visual Interval
    pH 8.6 Pink:
    Pass
  • Visual Interval
    pH 10.0 Red:
    Pass

 

 

Frequently Bought Together