Order Entry
Germany
ContactUsLinkComponent
Orthoboric acid 10 g/l in aqueous solution
Orthoboric acid 10 g/l in aqueous solution
Catalog # KRAF05918.6400
Supplier:  Bernd Kraft
CAS Number:  
undefined
Orthoboric acid 10 g/l in aqueous solution
Catalog # KRAF05918.6400
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337