Orthoboric acid 10 g/l in aqueous solution
Catalog # KRAF05735.7000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:20 L
- Seq:5302
Specifications
About this item
:
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337