Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ortho-Boric acid
Orthoboric acid 10 g/l in aqueous solution
Orthoboric acid 10 g/l in aqueous solution
Orthoboric acid 10 g/l in aqueous solution
KRAF05456.7000
:
Bernd Kraft
:
Orthoboric acid 10 g/l in aqueous solution
KRAF05456.7000
:
Bernd Kraft
:
Pack type:
Plastic bottle
Pk:
20 L
Seq:
5302
:
:
61.83 g/mol
:
Ambient
:
233-139-2
:
10043-35-3
:
B(OH)₃
:
MFCD00011337