Sieves, Monofilament Textile
: -
The monofilament textile comprises fat and even (monofile) fibres. The values for the monofilament textile openings are defined exactly and their measurements checked. The mono- filament textile is heat-set and this provides the mesh with its stability. It can hold defined particle sizes back on the cloth‘s surface.
- Abrasion resistance
- Fineness tolerance: ±10%
- Dimensions tolerance: ±10 mm
Sieving is basically grading defined particle sizes with an efective selectivity. We recommend monoflament textiles made of polyester for acids and monoflament textiles made of polyamide for bases. We can also provide polypropylene and many other materials upon request. We recommend our polyamide monoflament textile with carbon fibres as an antistatic option.