Order Entry
Germany
ContactUsLinkComponent
Hexafluorosilicic acid 34% in aqueous solution, Sigma-Aldrich®
Corrosive
Hexafluorosilicic acid 34% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL01302-1L
Supplier:  Merck
CAS Number:  
Corrosive
Hexafluorosilicic acid 34% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL01302-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :144.09 g/mol
Density:1.29…1.33 g/cm³ (20 °C)
UN:1778
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:241-034-8
 :GHS05
CAS Number:16961-83-4
Formula:H₂F₆Si
MDL Number:MFCD00036289