Order Entry
Germany
ContactUsLinkComponent
Hexafluorosilicic acid 25% (w/w) in aqueous solution
Corrosive
Hexafluorosilicic acid 25% (w/w) in aqueous solution
Certificates
undefined
Corrosive
Hexafluorosilicic acid 25% (w/w) in aqueous solution

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Assay:
    19 to 20.6 % (total fluorine)
  • GC:
    ≥94 %
  • Specific optical rotation:
    +24° to +28° (20°C, 589 nm) (C=1, Methanol)

Specification Test Results

Related Information

Frequently Bought Together