Sodium alginate (from Macrocystis pyrifera (kelp)), Bioreagent suitable for immobilisation of micro-organisms, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9005-38-3
:
MDL Number:MFCD00081310
Melting Pt:>300 °C
Storage Temperature:Ambient
Frequently Bought Together