Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Alginic acid
Alginic acid
Alginic acid
Alginic acid
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Alginic acid
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
9005-32-7
:
Formula:
(C₆H₈O₆)n
MDL Number:
MFCD00081309
:
176.13 g/mol
Merck Index:
14,242
Melting Pt:
>300 °C
Storage Temperature:
Ambient
EINECS:
232-680-1
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA17582
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation