Alginic acid
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9005-32-7
:
Formula:(C₆H₈O₆)n
MDL Number:MFCD00081309
:176.13 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:232-680-1
Specification Test Results
- Appearance:White to yellow to light beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15 % (2 g, 105°C, 4 hrs)
- Sulfated ash:≤8 %
- Titration excess NaOH:19 to 25 % (Carboxyl groups) On dried substance