Triamterene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:396-01-0
:
Formula:C₁₂H₁₁N₇
MDL Number:MFCD00006708
:253.27 g/mol
Merck Index:12,09731
Boiling Pt:573 °C (1013 hPa)
Melting Pt:316.1 °C
:M-Clarity™ quality level = MQ100
EINECS:206-904-3