
3-Bromostyrene 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2039-86-3
:
Formula:C₈H₇Br
MDL Number:MFCD00000088
:183.05 g/mol
Boiling Pt:74…75 °C (3 mmHg)
Density:1.406 g/cm³
Flash Pt:67 °C
EINECS:218-025-2
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Stabilizer:0.05 % 3,5-di-tert.-Butylcatechol