0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
2-Bromo-4,6-di-tert-butylphenol
2-Bromo-4,6-di-tert-butylphenol ≥97%
2-Bromo-4,6-di-tert-butylphenol ≥97%
:
Thermo Fisher Scientific
2-Bromo-4,6-di-tert-butylphenol ≥97%
:
Thermo Fisher Scientific
:
20834-61-1
:
:
C₁₄H₂₁BrO
:
MFCD00051597
:
285.22 g/mol
:
55…59 °C
:
GHS07
:
Ambient
msds_ALFAA13218
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years