Orders LinkContactUsLinkComponent
Plastic Box with White Lid
Plastic box with safety lock and white lid, 3 L, 193×193×112 mm, PP
  MULC25685
 :   MULTIROIR
Plastic Box with White Lid
  MULC25685
 :  MULTIROIR
Plastic Box with White Lid
Plastic Box with White Lid
Plastic box with safety lock and white lid, 3 L, 193×193×112 mm, PP
290,00 €
1 * 72 ST
Box opaque with airtight and tamper-evident lid, perfect for use in laboratories.
  • Polypropylene material
  • Opaque white
  • Watertight lid
  • Tamper-proof lid
  • Stackable box
  • Food contact quality
Ideal for laboratory environments, this versatile container ensures the secure storage and transport of samples and reagents. The airtight, tamper-evident lid provides enhanced safety, while the stackable design maximises space efficiency. The opaque white polypropylene construction ensures durability and easy identification of contents.
  • Capacity:
    3 L
  • Description:
    Plastic box with safety lock and white lid
  • Material:
    PP
  • W×D×H:
    193×193×112 mm