Order Entry
Germany
ContactUsLinkComponent
Ampicillin sodium salt ≥93%
Ampicillin sodium salt ≥93%
Catalog # ENZOALX380268G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ampicillin sodium salt ≥93%
Catalog # ENZOALX380268G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :371.39 g/mol
Storage Temperature:Freezer
CAS Number:69-52-3
Formula:C₁₆H₁₈N₃NaO₄S
MDL Number:MFCD00064313