Order Entry
Germany
ContactUsLinkComponent
Indirubin-3'-oxime ≥98%
Indirubin-3'-oxime ≥98%
Catalog # ENZOBMLCC2070005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Indirubin-3'-oxime ≥98%
Catalog # ENZOBMLCC2070005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :277.283 g/mol
Storage Temperature:Freezer
CAS Number:160807-49-8
Formula:C₁₆H₁₁N₃O₂
MDL Number:MFCD02683594