Order Entry
Germany
ContactUsLinkComponent
Nigericin sodium salt ≥98%
Nigericin sodium salt ≥98%
Catalog # ENZOBMLCA4210005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Nigericin sodium salt ≥98%
Catalog # ENZOBMLCA4210005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :746.94 g/mol
CAS Number:28643-80-3
 :MFCD27977549
Formula:C₄₀H₆₇NaO₁₁
MDL Number:MFCD00036825