Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methyladenine
3-Methyladenine ≥98% (by HPLC)
3-Methyladenine ≥98% (by HPLC)
ENZOBMLAP5020025
:
Enzo Life Sciences
:
3-Methyladenine ≥98% (by HPLC)
ENZOBMLAP5020025
:
Enzo Life Sciences
:
Pk:
25 mg
Seq:
2315
:
:
149.16 g/mol
:
5142-23-4
:
C₆H₇N₅
:
MFCD00010531