Order Entry
Germany
ContactUsLinkComponent
Sulindac sulphone ≥94% (by HPLC)
Sulindac sulphone ≥94% (by HPLC)
Catalog # ENZOBMLAP2010025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Sulindac sulphone ≥94% (by HPLC)
Catalog # ENZOBMLAP2010025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :372.4 g/mol
Storage Temperature:Ambient
CAS Number:59864-04-9
 :59973-80-7
Formula:C₂₀H₁₇FO₄S
MDL Number:MFCD00871143